Learn More
Medchemexpress LLC HY-122641B 5mg Medchemexpress, Deltasonamide 2 (TFA) CAS:2235358-74-2 Purity:>98%
SDP
Catalog No. 501932570
Encompass_Preferred
Medchemexpress, HY-122641B 5mg Deltasonamide 2 (TFA) CAS:2235358-74-2 Deltasonamide 2 TFA is competitive, high affinity PDEδ inhibitor with a K d of ~385 pM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2570 Supplier Medchemexpress LLC Supplier No. HY122641B5MG
Medchemexpress, HY-122641B 5mg Deltasonamide 2 (TFA) CAS:2235358-74-2 Deltasonamide 2 TFA is competitive, high affinity PDEδ inhibitor with a K d of ~385 pM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.