Learn More
Medchemexpress LLC HY-N2283 5mg Medchemexpress, Deltonin CAS:55659-75-1 Purity:>98%
SDP
Catalog No. 501933900
Encompass_Preferred
Medchemexpress, HY-N2283 5mg Deltonin CAS:55659-75-1 Deltonin, a steroidal saponin, isolated from Dioscorea zingiberensis Wright, with antitumor activity; Deltonin inhibits ERK1/2 and AKT activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3900 Supplier Medchemexpress LLC Supplier No. HYN22835MG
Medchemexpress, HY-N2283 5mg Deltonin CAS:55659-75-1 Deltonin, a steroidal saponin, isolated from Dioscorea zingiberensis Wright, with antitumor activity; Deltonin inhibits ERK1/2 and AKT activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.