Learn More
MSC Deltronic Class X Plus Pin Gage, 0.1755 in Diameter, 2-1/2 in Length
Catalog No. 5000511916
Encompass
This Deltronic Class X Plus Pin Gage is a precision tool designed for production inspection requirements. It features a 0.1755 inch diameter and a 2-1/2 inch length, constructed from durable tool steel with a tolerance of 0.00004 "
  • Precision lapped with approximately one micro-inch mirror finish.
  • Patented relief handle allows use with or without gage handles.
  • Provides absolute concentricity from end to end.
  • Inspection grade, round within 20 millionths.
  • Hardened and cold stabilized to Rockwell C 62-64 for extended life.
  • Includes a Certificate of Accuracy with NIST Traceability.
Catalog No. 50-005-11916 Supplier MSC Supplier No. 89567325
This Deltronic Class X Plus Pin Gage is a precision tool designed for production inspection requirements. It features a 0.1755 inch diameter and a 2-1/2 inch length, constructed from durable tool steel with a tolerance of 0.00004 "
  • Precision lapped with approximately one micro-inch mirror finish.
  • Patented relief handle allows use with or without gage handles.
  • Provides absolute concentricity from end to end.
  • Inspection grade, round within 20 millionths.
  • Hardened and cold stabilized to Rockwell C 62-64 for extended life.
  • Includes a Certificate of Accuracy with NIST Traceability.