Learn More
MSC Deltronic Class X Plus Pin Gage, 0.4540 in Diameter, 3 in Length, Tool Steel
Catalog No. 5000531656
Encompass
This pin gage is designed for precise measurement and inspection tasks, offering accurate qualification of hole sizes. Made from durable tool steel, it features a 0.4540 inch diameter and a 3 inch length, with a 4 Micro finish, making it ideal for production inspection. It can be used for determining Go/No-go tolerances or finding exact hole sizes, and comes with a Certificate of Accuracy with NIST Traceability.
  • Accuracy of plus 40 millionths, minus nothing.
  • Suitable for production inspection.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from tool steel.
  • Features a 4 micro finish.
  • Versatile for zeroing comparators, setting snap gages, checking micrometers, and more.
Catalog No. 50-005-31656 Supplier MSC Supplier No. 89595177
This pin gage is designed for precise measurement and inspection tasks, offering accurate qualification of hole sizes. Made from durable tool steel, it features a 0.4540 inch diameter and a 3 inch length, with a 4 Micro finish, making it ideal for production inspection. It can be used for determining Go/No-go tolerances or finding exact hole sizes, and comes with a Certificate of Accuracy with NIST Traceability.
  • Accuracy of plus 40 millionths, minus nothing.
  • Suitable for production inspection.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from tool steel.
  • Features a 4 micro finish.
  • Versatile for zeroing comparators, setting snap gages, checking micrometers, and more.