Learn More
MSC Deltronic Class X Plus Pin Gage, 0.4997 in Dia, 3 in Length, 4 Micro
Catalog No. 5000530457
Encompass
This Class X Plus Pin Gage is designed for precise production inspection requirements. It features a 0.4997 in diameter and a 3 in length, constructed from durable tool steel with a 4 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for precise production inspection requirements.
  • Constructed from durable tool steel.
  • Features a 4 micro coating/finish.
  • Includes a Certificate of Accuracy with NIST Traceability.
Catalog No. 50-005-30457 Supplier MSC Supplier No. 89599740
This Class X Plus Pin Gage is designed for precise production inspection requirements. It features a 0.4997 in diameter and a 3 in length, constructed from durable tool steel with a 4 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for precise production inspection requirements.
  • Constructed from durable tool steel.
  • Features a 4 micro coating/finish.
  • Includes a Certificate of Accuracy with NIST Traceability.