Learn More
MSC Deltronic Double Reversible & Single Reversible Plug Gage Handle Bushing, 0.152-0.159 inch
Catalog No. 5000510488
Encompass
The Deltronic Double Reversible & Single Reversible Plug Gage Handle Bushing is designed for use in production inspection applications. Its reversible design offers versatility, allowing for both double and single reversible configurations. The bushing accommodates measurements from 0.1520 to 0.1590 decimal inches. Manufactured in the United States, this component ensures quality and adherence to specific measurement standards for plug gages.
  • Reversible design offers versatility
  • Accommodates measurements from 0.152 to 0.159 inch
  • Ensures quality and adherence to specific measurement standards
  • Designed for production inspection applications
  • Manufactured in the United States
Catalog No. 50-005-10488 Supplier MSC Supplier No. 72729197
The Deltronic Double Reversible & Single Reversible Plug Gage Handle Bushing is designed for use in production inspection applications. Its reversible design offers versatility, allowing for both double and single reversible configurations. The bushing accommodates measurements from 0.1520 to 0.1590 decimal inches. Manufactured in the United States, this component ensures quality and adherence to specific measurement standards for plug gages.
  • Reversible design offers versatility
  • Accommodates measurements from 0.152 to 0.159 inch
  • Ensures quality and adherence to specific measurement standards
  • Designed for production inspection applications
  • Manufactured in the United States