Learn More
MSC Deltronic Pin Gage, 0.0319 in Diameter, 2-1/4 in Length, Tool Steel, Class X Plus
Catalog No. 5000527631
Encompass
Deltronic Class X Plus Pin Gages are engineered for precise production inspection. These durable tool steel gages feature a 0.0319 in diameter and a 2-1/4 in length, ensuring accurate measurements. Each gage includes a 4 micro coated finish and a Certificate of Accuracy with NIST Traceability for reliable performance.
  • Engineered for production inspection requirements
  • Constructed from tool steel
  • Measures 0.0319 in in diameter
  • Features a 2-1/4 in length
  • Includes a 4 micro coated finish
  • Comes with a certificate of accuracy with NIST traceability
  • Designed with plus tolerance
Catalog No. 50-005-27631 Supplier MSC Supplier No. 86300001
Deltronic Class X Plus Pin Gages are engineered for precise production inspection. These durable tool steel gages feature a 0.0319 in diameter and a 2-1/4 in length, ensuring accurate measurements. Each gage includes a 4 micro coated finish and a Certificate of Accuracy with NIST Traceability for reliable performance.
  • Engineered for production inspection requirements
  • Constructed from tool steel
  • Measures 0.0319 in in diameter
  • Features a 2-1/4 in length
  • Includes a 4 micro coated finish
  • Comes with a certificate of accuracy with NIST traceability
  • Designed with plus tolerance