Learn More
MSC Deltronic Pin Gage, 0.4994 in Diameter, Class X, Tool Steel
Catalog No. 5000536499
Encompass
This Class X pin gage is designed for precise production inspection requirements. Manufactured from hardened and cold-stabilized tool steel, it offers extended life and features a precision-lapped surface with a mirror finish, ensuring high accuracy and absolute concentricity.
  • Diameter of 0.4994 "
  • Tolerance of 0.00004 "
  • Constructed from hardened and cold stabilized tool steel for extended life (Rockwell C 62-64).
  • Precision lapped surface with a 1 micro-inch mirror finish.
  • Round within 20 millionths.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a patented relief handle for versatile use.
  • Absolute concentricity from end to end.
Catalog No. 50-005-36499 Supplier MSC Supplier No. 89599716
This Class X pin gage is designed for precise production inspection requirements. Manufactured from hardened and cold-stabilized tool steel, it offers extended life and features a precision-lapped surface with a mirror finish, ensuring high accuracy and absolute concentricity.
  • Diameter of 0.4994 "
  • Tolerance of 0.00004 "
  • Constructed from hardened and cold stabilized tool steel for extended life (Rockwell C 62-64).
  • Precision lapped surface with a 1 micro-inch mirror finish.
  • Round within 20 millionths.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a patented relief handle for versatile use.
  • Absolute concentricity from end to end.