Learn More
MSC Deltronic Pin Gage, Class X, .1270 Inch
Catalog No. 5000530456
Encompass
The Class X Pin Gage is a precision measuring tool designed for accurate gauging applications. This specific gage has a diameter of 0.1270 inches.
  • Precision tool for accurate gauging
  • Class X accuracy for high reliability
  • Diameter of 0.1270 inches
  • Length of 2 1/2 inches (63.50 mm)
Catalog No. 50-005-30456 Supplier MSC Supplier No. 89562474
The Class X Pin Gage is a precision measuring tool designed for accurate gauging applications. This specific gage has a diameter of 0.1270 inches.
  • Precision tool for accurate gauging
  • Class X accuracy for high reliability
  • Diameter of 0.1270 inches
  • Length of 2 1/2 inches (63.50 mm)