Learn More
MSC Deltronic Pin Gage, Class X Plus, 0.0331 in, 2-1/4 in
Catalog No. 5000531298
Encompass
The Deltronic Class X Plus Pin Gage is designed for precise production inspection requirements. It features a 0.0331 inch diameter and a 2-1/4 inch length, made from durable Tool Steel. Each gage comes with a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements.
  • Class X Plus Pin Gage.
  • 4 Micro Coated finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from tool steel.
  • Designed for production inspection requirements.
  • Country of origin: United States.
Catalog No. 50-005-31298 Supplier MSC Supplier No. 86300126
The Deltronic Class X Plus Pin Gage is designed for precise production inspection requirements. It features a 0.0331 inch diameter and a 2-1/4 inch length, made from durable Tool Steel. Each gage comes with a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements.
  • Class X Plus Pin Gage.
  • 4 Micro Coated finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from tool steel.
  • Designed for production inspection requirements.
  • Country of origin: United States.