Learn More
MSC Deltronic Pin Gage, Class X Plus, 0.5860 in, 3 in, Tool Steel
Catalog No. 5000530798
Encompass
This Class X Plus Pin Gage is designed for precise production inspection requirements. Made from durable tool steel, it features a 0.5860 inch diameter and a 3 inch length, ensuring high accuracy for critical measurements.
  • Certificate of accuracy with NIST traceability included.
  • Made from robust tool steel for durability.
  • 0.5860 inch diameter for precise measurements.
  • 3 inch length for versatile applications.
  • Features a 4 micro finish for enhanced performance.
  • Suitable for production inspection requirements.
Catalog No. 50-005-30798 Supplier MSC Supplier No. 72686017
This Class X Plus Pin Gage is designed for precise production inspection requirements. Made from durable tool steel, it features a 0.5860 inch diameter and a 3 inch length, ensuring high accuracy for critical measurements.
  • Certificate of accuracy with NIST traceability included.
  • Made from robust tool steel for durability.
  • 0.5860 inch diameter for precise measurements.
  • 3 inch length for versatile applications.
  • Features a 4 micro finish for enhanced performance.
  • Suitable for production inspection requirements.