Learn More
Cayman Chemical DeltrphIn CtrIfluoroACTAt 10mg
SDP
Catalog No. 502989930
Encompass_Preferred
A peptide agonist of δ-opioid receptors (Ki = 0.15 nM); selective for δ-opioid receptors over µ- and ĸ-opioid receptors; induces δ-opioid receptor translocation from large dense-core vesicles to the cell surface and CGRP release in isolated mouse DRG cells at 0.1 and 1 µM, respectively; increases latency to withdraw in the paw pressure and tail-flick tests in rats at 0.15, 1.5, and 15 UG/animal intrathecally
Catalog No. 50-298-9930 Supplier Cayman Chemical Supplier No. 3005010 MG
A peptide agonist of δ-opioid receptors (Ki = 0.15 nM); selective for δ-opioid receptors over µ- and ĸ-opioid receptors; induces δ-opioid receptor translocation from large dense-core vesicles to the cell surface and CGRP release in isolated mouse DRG cells at 0.1 and 1 µM, respectively; increases latency to withdraw in the paw pressure and tail-flick tests in rats at 0.15, 1.5, and 15 UG/animal intrathecally