Learn More
TARGETMOL CHEMICALS INC Delucemine Hydrochloride 1MG
SDP
Catalog No. 502878224
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Delucemine Hydrochloride (Delucemine) is a polyamine NMDA receptor antagonist used in the study of neurological disorders such as depression and Alzheimer's disease. Purity 99.53%
Catalog No. 50-287-8224 Supplier TARGETMOL CHEMICALS INC Supplier No. T271431MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Delucemine Hydrochloride (Delucemine) is a polyamine NMDA receptor antagonist used in the study of neurological disorders such as depression and Alzheimer's disease. Purity 99.53%