Learn More
Description
- Demonstrate proper condom use with this realistic training model made of lifelike BIOLIKE 2™ synthetic tissue.
- The model 'ejaculates' UV light-sensitive fluid, which illuminates under a UV flashlight, and helps demonstrate the importance of avoiding contact with body fluids during intercourse.
- A great resource to educate any group on the important role condoms can play in preventing STIs and unplanned pregnancies. Also a perfect Personal, Social, Health and Economic (PSHE) education teaching tool for schools and health education professionals.
- Comes with syringe, UV light-sensitive fluid, 12 condoms, and carrying case.
- UV flashlight not included.
- This model comes with condoms made from latex.
Specifications
Specifications
| Product Type | Condom Training Model |
| For Use With (Application) | Personal, Social, Health and Economic (PSHE) Education |
| Color | White Skin Tone |
| Dimensions (L x W x H) | 13 x 24 x 13 cm |
| For Use With (Equipment) | UV Flashlight |
| Gender | Male |
| Includes | Syringe, UV Light-Sensitive Fluid, 12 Condoms, Carrying Case |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


