Learn More
3B Scientific™ Deluxe Demonstration Human Skull with 3B Smart Anatomy
Shop All 3B Scientific Products
Description
Features:
- On the left half, the temporal bone can be removed and folded up in the area of the tympanic membrane.
- Maxilla and mandible of the skull are opened to reveal the alveolar nerves.
- On the right side the temporal bone is opened to reveal the sigmoid sinus, the facial nerve canal and the semicircular ducts.
- Additional flaps on the skull are located at the maxillary sinus and the right half of the mandible, so that the dental roots of the premolars and molars of the lower jaw can also be viewed.
- Natural occlusion and the individual removal and replacement of each tooth make this skull especially interesting for dentists.
- 3B Smart Anatomy course includes:
- 23 digital anatomy lectures
- 117 different virtual anatomy models
- 39 anatomy quizzes
- Warranty upgrade from 3 to 5 years with every product registration
- Access to a free 3-day trial to all premium features of the Complete Anatomy app
Specifications
Specifications
| For Use With (Application) | Anatomy studies |
| Includes | 10 removable parts; Smart Anatomy software |
| Product Type | Skull, Demonstration |