Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC MetroVac Deluxe Cordless Hand Vac, Black, 250W, Dual Battery

Catalog No. 502836234
Encompass

This cordless hand vacuum delivers professional-grade cleaning with its 250W BLDC motor, providing nearly 100 inches of water lift for powerful dry pick-up. The deluxe model features a dual interchangeable 22.2V lithium-ion battery system and advanced HEPA filtration, ensuring extended use and efficient cleaning. Its lightweight design and multiple power levels make it ideal for various cleaning tasks, from car interiors to small spaces.

  • Three power levels: Low, Medium, High
  • Lightweight and easy to maneuver
  • 250W motor for deep-cleaning performance
  • Battery level indicator lights
  • All steel construction with black matte finish
  • Dual interchangeable 22.2V/2000mAH battery packs
  • HEPA filter and filter bag included
  • Comes with a 3' hose, upholstery nozzle, crevice tool, dust brush, and shoulder strap

Catalog No. 50-283-6234 Supplier MSC Supplier No. 59379636
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This cordless hand vacuum delivers professional-grade cleaning with its 250W BLDC motor, providing nearly 100 inches of water lift for powerful dry pick-up. The deluxe model features a dual interchangeable 22.2V lithium-ion battery system and advanced HEPA filtration, ensuring extended use and efficient cleaning. Its lightweight design and multiple power levels make it ideal for various cleaning tasks, from car interiors to small spaces.

  • Three power levels: Low, Medium, High
  • Lightweight and easy to maneuver
  • 250W motor for deep-cleaning performance
  • Battery level indicator lights
  • All steel construction with black matte finish
  • Dual interchangeable 22.2V/2000mAH battery packs
  • HEPA filter and filter bag included
  • Comes with a 3' hose, upholstery nozzle, crevice tool, dust brush, and shoulder strap

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.