Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Spunbond Polypropylene Lab Coats, Knit Wrists and Collar

Catalog No. 19304127 Shop All Cellucap Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-304127 2X-Large
19-304126 X-Large
19-304125 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-304127 Supplier Cellucap Supplier No. 3330WXXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lab coats with a heavier polypropylene alternative for splash protection from non-hazardous liquids.

  • Lab coats made with heavier 1.75 oz spunbond polypropylene
  • Provides non-hazardous splash and dry particulate protection
  • Snap front
  • Packaged 25 per case
  • Three total pockets, one breast and two hip
  • White, knit cuffs, knit collar

Specifications

Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Spunbond Polypropylene
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Collar Type Knit
Model 3330W
For Use With (Application) For splash protection from non-hazardous liquids and dry particulates
Weight (English) 0.06 lb.
Weight (Metric) 0.03 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.