Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Deluxe Owl Pellet Dissection Lab Activity
GREENER_CHOICESCI_ED

Catalog No. s29557 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29557 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Lead and Lead Compounds, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S29557 Supplier Innovating Science™ Supplier No. IS3016
Only null left
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Lead and Lead Compounds, which are known to the State of California to cause cancer and reproductive harm.

Innovating Science™ Deluxe Owl Pellet Dissection Lab Activity can teach students the concepts of food webs and food chains while identifying the small animals that are prey for the owls.

  • Owls have a specialized digestive tract which helps them expel the indigestible parts of their prey in pellet form
  • The pellets are heat sterilized and individually wrapped
  • Kit Includes:
    • 15 Owl Pellets
    • 30 Dissecting Probes
    • 15 Forceps
    • 15 Plastic Rulers
    • 15 Hand Magnifiers
    • 60 Nitrile Gloves

Specifications

For Use With (Application) Biology Class
Includes Teachers Manual and Student Guide
Size Deluxe
Length (English) 9.75 in.
Class Size 15 Lab Groups
Product Type Owl Pellet Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.