Learn More
Research Products International Corp Deluxe PCR Workstation, Assorted Colors, 5 per Case
SDP
Catalog No. 50212575
Encompass
  • Ideal for all PCR applications.
  • Sturdy, Reusable, Autoclavable.
  • Removable Slide Out PCR rack for added workspace flexibility.
This "Deluxe" PCR Workstation system provides convenient handling for a variety of tubes used in pre and post PCR applications. 
Create a personnal workspace with removable, slide-out PCR rack for added benchtop flexibility. 
Workstation consists of base rack, removable slide-out rack and lid. Features alpha/numerical index. 
Slide-out PCR Rack holds 96 PCR tubes. Base rack holds 32x0.2ml: 24x1.5-2.0ml: and 16x0.5ml. SKU 248651 is supplied as Single PCR Workstation with single base rack and slide-out PCR rack, choose individual color.
Catalog No. 50-212-575 Supplier Research Products International Corp Supplier No. 248640
  • Ideal for all PCR applications.
  • Sturdy, Reusable, Autoclavable.
  • Removable Slide Out PCR rack for added workspace flexibility.
This "Deluxe" PCR Workstation system provides convenient handling for a variety of tubes used in pre and post PCR applications. 
Create a personnal workspace with removable, slide-out PCR rack for added benchtop flexibility. 
Workstation consists of base rack, removable slide-out rack and lid. Features alpha/numerical index. 
Slide-out PCR Rack holds 96 PCR tubes. Base rack holds 32x0.2ml: 24x1.5-2.0ml: and 16x0.5ml. SKU 248651 is supplied as Single PCR Workstation with single base rack and slide-out PCR rack, choose individual color.