Learn More
Uline Deluxe Safety Tape 2 Rolls
Catalog No. NC2117612
Encompass
Orange safety tape in 2-inch width and 100-foot rolls with pressure-sensitive adhesive backing for surface application, meets ANSI and OSHA standards for hazard warning and restricted area delineation using bright orange color and retroreflective striping for visibility.
This item is not returnable. View return policy
Catalog No. NC2117612 Supplier Uline Supplier No. DELUXE SAFETY TAPE 2X100
Orange safety tape in 2-inch width and 100-foot rolls with pressure-sensitive adhesive backing for surface application, meets ANSI and OSHA standards for hazard warning and restricted area delineation using bright orange color and retroreflective striping for visibility.