Learn More
Perkin Elmer US LLC Deluxe Spares Kit for SC-FAST and oneFAST
Catalog No. 502105165
Encompass_Preferred
This FAST Deluxe Spares Kit includes the contents of the Standard Spares Kit plus a PFA-ST nebulizer and spare P7 FAST valve.
Catalog No. 50-210-5165 Supplier Perkin Elmer US LLC Supplier No. N0777432
Add to Cart
Add to Cart
Request a Quote
This FAST Deluxe Spares Kit includes the contents of the Standard Spares Kit plus a PFA-ST nebulizer and spare P7 FAST valve.