Learn More
Brady™ Deluxe Visitor Spec Dispenser
Catalog No. 19129478 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Clear
Fluorescent Green
Fluorescent Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-129-478 Fluorescent Orange
19-129-474 Clear
19-129-475 Fluorescent Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-129-478 Supplier Brady™ Supplier No. MVSDO
Add to Cart
Add to Cart
Eyewear dispensers remind employees to wear protective equipment and save much-needed room by storing the equipment in space-efficient containers.
  • With bottom door that keeps glasses in and folds down for easy dispensing
  • Each dispenser holds approximately 20 to 25 glasses
  • Dispensers are supplied with two top holes making them ideal for wall mounting
Specifications
Color Fluorescent Orange
Product Type Safety Glasses Dispenser (Unfilled)
Size (H x W x D) 18 x 8 x 4 in.
Material Acrylic