Learn More
Nelson Jameson Company DELVO TEST P MULTI 5PK
Catalog No. NC9617231
Encompass
This Delvotest P Multi 5 pack is a very economical option for NCIMS-compliant customers. It requires a dry bath and a 100 uL pipettor. For NCIMS use.
Catalog No. NC9617231 Supplier Nelson Jameson Company Supplier No. 2333480 RFC
This Delvotest P Multi 5 pack is a very economical option for NCIMS-compliant customers. It requires a dry bath and a 100 uL pipettor. For NCIMS use.