Learn More
Nelson Jameson Company DELVOTEST P TEST KIT
Catalog No. NC0924882
Encompass
The original version of this popular broad spectrum milk antibiotic test. Results are available in 2-1/2 hours. For NCIMS use. A simple, reliable, and economical test for antibiotic residues in milk. Delvotest detects a broad spectrum of antibiotics-not limited to the penicillin family. Designed for use on the farm or in a laboratory, this standard diffusion test has high sensitivity to most veterinary antibiotics. Results are indicated by color. Self-contained, except for the required block heater, each kit contains individual plastic ampules containing Bacillus stearothermophilus in a solid medium, nutrient tablets(not required for SP-NT), dosing syringe, disposable pipets, tweezers, and instructions.
Catalog No. NC0924882 Supplier Nelson Jameson Company Supplier No. 2333006 BFC
The original version of this popular broad spectrum milk antibiotic test. Results are available in 2-1/2 hours. For NCIMS use. A simple, reliable, and economical test for antibiotic residues in milk. Delvotest detects a broad spectrum of antibiotics-not limited to the penicillin family. Designed for use on the farm or in a laboratory, this standard diffusion test has high sensitivity to most veterinary antibiotics. Results are indicated by color. Self-contained, except for the required block heater, each kit contains individual plastic ampules containing Bacillus stearothermophilus in a solid medium, nutrient tablets(not required for SP-NT), dosing syringe, disposable pipets, tweezers, and instructions.