Learn More
ERA WATERS CORP DEMAND WASTEWATER QC STANDARD
SDP
Catalog No. NC9419096
Encompass
One 15 mL screw-cap vial yields up to 2 liters after dilution.
Catalog No. NC9419096 Supplier ERA WATERS CORP Supplier No. 516
One 15 mL screw-cap vial yields up to 2 liters after dilution.