Learn More
TARGETMOL CHEMICALS INC DEMECARIUM BROMIDE 50MG
SDP
Catalog No. 502579239
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Demecarium bromide (BC-48) is a potent cholinesterase inhibitorand is treatment glaucoma agent. purity: 98%
Catalog No. 50-257-9239 Supplier TARGETMOL CHEMICALS INC Supplier No. T721350MG
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Demecarium bromide (BC-48) is a potent cholinesterase inhibitorand is treatment glaucoma agent. purity: 98%