Learn More
Medchemexpress LLC Demecarium bromide | 56-94-0 | 95.0% | 1 ML
SDP
Catalog No. 5000412721
Encompass_Preferred
Demecarium Bromide (BC-48) is a potent cholinesterase inhibitor known for its apparent affinity (Kiapp) of 0.15 μM. It serves as a glaucoma agent, effective in both normotensive and inherited glaucoma conditions. In vivo studies demonstrate its ability to cause long-term miosis and decrease intraocular pressure.
  • Potent cholinesterase inhibitor
  • Apparent affinity (Kiapp) of 0.15 μM
  • Used as a glaucoma agent
  • Causes long-term miosis
  • Decreases intraocular pressure (IOP)
Catalog No. 50-004-12721 Supplier Medchemexpress LLC Supplier No. HYB1626A10MM/1ML
Demecarium Bromide (BC-48) is a potent cholinesterase inhibitor known for its apparent affinity (Kiapp) of 0.15 μM. It serves as a glaucoma agent, effective in both normotensive and inherited glaucoma conditions. In vivo studies demonstrate its ability to cause long-term miosis and decrease intraocular pressure.
  • Potent cholinesterase inhibitor
  • Apparent affinity (Kiapp) of 0.15 μM
  • Used as a glaucoma agent
  • Causes long-term miosis
  • Decreases intraocular pressure (IOP)