Learn More
Medchemexpress LLC Demecarium Bromide | 56-94-0 | 95.0% | 25 MG
SDP
Catalog No. 5000414051
Encompass_Preferred
Demecarium Bromide is described as a potent cholinesterase inhibitor, with an apparent affinity (Kiapp) of 0.15 μM. It is also used as a glaucoma agent.
  • Potent cholinesterase inhibitor
  • Apparent affinity (Kiapp) of 0.15 μM
  • Used as a glaucoma agent
Catalog No. 50-004-14051 Supplier Medchemexpress LLC Supplier No. 25MGHYB1626A
Demecarium Bromide is described as a potent cholinesterase inhibitor, with an apparent affinity (Kiapp) of 0.15 μM. It is also used as a glaucoma agent.
  • Potent cholinesterase inhibitor
  • Apparent affinity (Kiapp) of 0.15 μM
  • Used as a glaucoma agent