Learn More
Medchemexpress LLC Demecarium bromide | 56-94-0 | MFCD00152376 | 95.0% | 716.59 | C32H52Br2N4O4 | 10 MG
SDP
Catalog No. 5000297280
Encompass_Preferred
Demecarium bromide is a long-acting quaternary ammonium cholinesterase inhibitor used in research on glaucoma and cholinesterase activity. Supplied as the bromide salt, it has a reported apparent affinity (Kiapp) of approximately 0.15 μM and is provided with a product data sheet and SDS for safe handling.
  • Long-acting cholinesterase inhibitor for glaucoma research
  • Reported Kiapp ~0.15 μM against cholinesterase
  • High chemical purity (~95.0%) suitable for research use
  • Available in small quantities for laboratory experiments
  • Provided with a product data sheet and safety data sheet
Catalog No. 50-002-97280 Supplier Medchemexpress LLC Supplier No. HYB1626A10MG
Demecarium bromide is a long-acting quaternary ammonium cholinesterase inhibitor used in research on glaucoma and cholinesterase activity. Supplied as the bromide salt, it has a reported apparent affinity (Kiapp) of approximately 0.15 μM and is provided with a product data sheet and SDS for safe handling.
  • Long-acting cholinesterase inhibitor for glaucoma research
  • Reported Kiapp ~0.15 μM against cholinesterase
  • High chemical purity (~95.0%) suitable for research use
  • Available in small quantities for laboratory experiments
  • Provided with a product data sheet and safety data sheet