Learn More
Medchemexpress LLC HY-B1626A 5mg Medchemexpress, Demecarium Bromide CAS:56-94-0 Purity:>98%
SDP
Catalog No. 501932981
Encompass_Preferred
Medchemexpress, HY-B1626A 5mg Demecarium Bromide CAS:56-94-0 Demecarium Bromide (BC-48) is a potent cholinesterase inhibitor, with an apparent affinity ( K iapp ) of 0.15 μM [1] . Demecarium Bromide (BC-48) is used as a glaucoma agent [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2981 Supplier Medchemexpress LLC Supplier No. HYB1626A5MG
Medchemexpress, HY-B1626A 5mg Demecarium Bromide CAS:56-94-0 Demecarium Bromide (BC-48) is a potent cholinesterase inhibitor, with an apparent affinity ( K iapp ) of 0.15 μM [1] . Demecarium Bromide (BC-48) is used as a glaucoma agent [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.