Learn More
Apexbio Technology LLC Demeclocycline-HCl
SDP
Catalog No. 5000569391
Encompass_Preferred
Demeclocycline hydrochloride (CAS No 64-73-3) is a tetracycline antibiotic primarily used in biomedical research for its ability to modulate bacterial ribosome function thereby inhibiting protein synthesis Originating from Streptomyces aureofaciens its mechanism involves the blockade of the 30S ribosomal subunit preventing aminoacyl-tRNA attachment Commonly demonstrating in vitro activity in the nanomolar to low micromolar range this compound is utilized in cell models and animal studies to investigate bacterial infections and as a tool for drug screening Experimental concentrations typically depend on the specific research design allowing for versatility in various biological and pharmacological studies
Catalog No. 50-005-69391 Supplier Apexbio Technology LLC Supplier No. B3601500
Demeclocycline hydrochloride (CAS No 64-73-3) is a tetracycline antibiotic primarily used in biomedical research for its ability to modulate bacterial ribosome function thereby inhibiting protein synthesis Originating from Streptomyces aureofaciens its mechanism involves the blockade of the 30S ribosomal subunit preventing aminoacyl-tRNA attachment Commonly demonstrating in vitro activity in the nanomolar to low micromolar range this compound is utilized in cell models and animal studies to investigate bacterial infections and as a tool for drug screening Experimental concentrations typically depend on the specific research design allowing for versatility in various biological and pharmacological studies