Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Demeclocycline hydrochloride

Catalog No. AAJ6310203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6310203 1 g
AAJ63102MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6310203 Supplier Thermo Scientific Chemicals Supplier No. J6310203
Add to Cart

CAS: 64-73-3 | C21H22Cl2N2O8 | 501.313 g/mol

Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Demeclocycline hydrochloride is used as an antibiotic related to tetracycline and produced by streptomyces aureofaciens, and used to maintain effective blood levels for longer periods of time, used as a fluorescent bone-labeling agent in bone histomorphometry.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-73-3
Molecular Formula C21H22Cl2N2O8
Molecular Weight (g/mol) 501.313
MDL Number MFCD00082371
InChI Key OAPVUSSHCBRCOL-KBHRXELFSA-N
Synonym demeclocycline hydrochloride, demeclocycline hcl, declomycin, detravis, ledermycine, clortetrin, ledermycin, ledermicina, veraciclina, demeplus
PubChem CID 54686764
IUPAC Name (4S,4aS,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES CN(C)C1C2CC3C(C4=C(C=CC(=C4C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O)Cl)O.Cl

Specifications

Melting Point >245°C (decomposition)
Color Yellow
Quantity 1 g
Beilstein 3849198
Merck Index 14,2886
Solubility Information Soluble in water.
Formula Weight 501.32
Physical Form Powder
Chemical Name or Material Demeclocycline hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-592-2
RTECSNumber QI7700000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.