Learn More
Medchemexpress LLC Demecolcine | 477-30-5 | MFCD00075459 | 99.9% | 371.43 | C21H25NO5 | 10 MG
SDP
Catalog No. 5000296531
Encompass_Preferred
Demecolcine is a colchicine-derivative mitotic inhibitor used in research to disrupt microtubule polymerization and to study cell division, apoptosis, and cytoskeleton-related pathways. It is supplied as a research-grade small molecule with defined storage recommendations for powder and solution states.
  • Potent tubulin inhibitor (IC50 ≈ 2.4 μM).
  • Useful for cell cycle and apoptosis studies.
  • Available in small research pack sizes (e.g., 10 MG).
  • Powder stable at -20°C for extended storage; solution stability specified for frozen storage.
  • Datasheet provides CAS, formula, molecular weight, and handling guidance.
Catalog No. 50-002-96531 Supplier Medchemexpress LLC Supplier No. HYN028210MG
Demecolcine is a colchicine-derivative mitotic inhibitor used in research to disrupt microtubule polymerization and to study cell division, apoptosis, and cytoskeleton-related pathways. It is supplied as a research-grade small molecule with defined storage recommendations for powder and solution states.
  • Potent tubulin inhibitor (IC50 ≈ 2.4 μM).
  • Useful for cell cycle and apoptosis studies.
  • Available in small research pack sizes (e.g., 10 MG).
  • Powder stable at -20°C for extended storage; solution stability specified for frozen storage.
  • Datasheet provides CAS, formula, molecular weight, and handling guidance.