Learn More
Cayman Chemical Demethoxycurcumin, Purity: ≥98%, CAS: 22608-11-3, 10 mg
SDP
Catalog No. 502951935
Encompass_Preferred
A natural demethoxy derivative of curcumin; suppresses proliferation in cancer cells at 50-100 μM; down-regulates p300 and inhibits LPS-induced iNOS expression
Catalog No. 50-295-1935 Supplier Cayman Chemical Supplier No. 1096110 MG
A natural demethoxy derivative of curcumin; suppresses proliferation in cancer cells at 50-100 μM; down-regulates p300 and inhibits LPS-induced iNOS expression