Learn More
AdipoGen Demethoxycurcumin (5 mg)
SDP
Supplier:  AdipoGen CDXD0929M005
Encompass_Preferred
Chemical. CAS: 22608-11-3. Formula: C20H18O5. MW: 338.35. Demethoxycurcumin (DMC) is a natural demethoxy derivative of curcumin with anti-inflammatory and anti-cancer properties. DMC suppresses cell proliferation, migration and invasion in cancer cells. It down-regulates the transcriptional coactivator p300, suppressing the Wnt/beta-catenin pathway, and inhibits lipopolysaccharide induction of iNOS by blocking NF-kappa activation. It also has been shown to inhibit energy metabolic and oncogenic signaling pathways through AMPK activation. In addition it is a potent inhibitor of P-Type ATPases. Shown to have neuroprotective properties. It also has anti-plasmodial activity.
Catalog No. 50-205-4409