Learn More
Medchemexpress LLC Demethoxyencecalin | 19013-07-1 | 99.3% | C13H14O2 | 100 MG
SDP
Catalog No. 5000433184
Encompass_Preferred
Demethoxyencecalin is a chromene isolated from Helianthus annuus, known for its antifungal activities. It is intended for research use only.
  • Exhibits antifungal activities
  • Isolated from Helianthus annuus
  • For research use only
  • Molecular weight of 202.25
  • Appears as a yellow to brown liquid
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-33184 Supplier Medchemexpress LLC Supplier No. HY77173100MG
Demethoxyencecalin is a chromene isolated from Helianthus annuus, known for its antifungal activities. It is intended for research use only.
  • Exhibits antifungal activities
  • Isolated from Helianthus annuus
  • For research use only
  • Molecular weight of 202.25
  • Appears as a yellow to brown liquid
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month