Learn More
Selleck Chemical LLC Demethyl-Coclaurine S3294-5mg
SDP
Catalog No. 503108783
Encompass_Preferred
Demethyl-Coclaurine (Higenamine Norcoclaurine) the key component of the Chinese herb aconite root is a beta-2 adrenergic receptor ( 2-AR) agonist Demethyl-Coclaurine stimulates AKT phosphorylation and requires PI3K activation for the anti-apoptotic effect in cardiomyocytes
Catalog No. 50-310-8783 Supplier Selleck Chemical LLC Supplier No. S32945MG
Demethyl-Coclaurine (Higenamine Norcoclaurine) the key component of the Chinese herb aconite root is a beta-2 adrenergic receptor ( 2-AR) agonist Demethyl-Coclaurine stimulates AKT phosphorylation and requires PI3K activation for the anti-apoptotic effect in cardiomyocytes