Learn More
Cayman Chemical Demethylwedelolacton 10mg
SDP
Catalog No. 502951971
Encompass_Preferred
A coumestan with diverse biological activities; reduces CCL4- and GaIN-induced cytotoxicity in rat hepatocytes in a dose-dependent manner; inhibits lethal and hemorrhagic activities of C. rhodostoma venom; inhibits anchorage-independent cell growth of MDA-MB-231 breast cancer cells; decreases the number of lung metastases in an MDA-MB-231 xenograft model in nude mice
Catalog No. 50-295-1971 Supplier Cayman Chemical Supplier No. 1170310 MG
A coumestan with diverse biological activities; reduces CCL4- and GaIN-induced cytotoxicity in rat hepatocytes in a dose-dependent manner; inhibits lethal and hemorrhagic activities of C. rhodostoma venom; inhibits anchorage-independent cell growth of MDA-MB-231 breast cancer cells; decreases the number of lung metastases in an MDA-MB-231 xenograft model in nude mice