Learn More
Medchemexpress LLC Demethylwedelolactone | 6468-55-9 | 300.22 | 5 MG
SDP
Catalog No. 5000415845
Encompass_Preferred
Demethylwedelolactone is a naturally occurring coumestan isolated from *Eclipta alba*. It is a potent trypsin inhibitor with an IC50 of 3.0 μM. This compound has been observed to suppress cell motility and cell invasion in breast cancer cells.
  • Naturally occurring coumestan isolated from *Eclipta alba*.
  • Potent trypsin inhibitor (IC50: 3.0 μM).
  • Suppresses cell motility and cell invasion of breast cancer cells.
  • Purity: 99.89%.
Catalog No. 50-004-15845 Supplier Medchemexpress LLC Supplier No. HYN20285MG
Demethylwedelolactone is a naturally occurring coumestan isolated from *Eclipta alba*. It is a potent trypsin inhibitor with an IC50 of 3.0 μM. This compound has been observed to suppress cell motility and cell invasion in breast cancer cells.
  • Naturally occurring coumestan isolated from *Eclipta alba*.
  • Potent trypsin inhibitor (IC50: 3.0 μM).
  • Suppresses cell motility and cell invasion of breast cancer cells.
  • Purity: 99.89%.