Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Demethylzeylasteral | 107316-88-1 | MFCD16660658 | 99.9% | 480.59 | C29H36O6 | 10 MG
SDP

Catalog No. 5000296427
Encompass_Preferred

Demethylzeylasteral is an orally active triterpenoid compound isolated from Tripterygium wilfordii that functions as an apoptosis inducer and has reported anti-inflammatory and antitumor activities. Supplied as a high-purity solid for research applications, it is suitable for use in biochemical and cell-based studies when prepared according to laboratory protocols.

  • Apoptosis inducer with reported anti-inflammatory and antitumor activity.
  • CAS number 107316-88-1 for unambiguous identification.
  • Molecular formula C29H36O6; molecular weight 480.59.
  • High purity (≈99.9%) in solid form, suitable for experimental work.
  • Available in multiple sizes, including 10 mg and millimolar DMSO solutions.
  • Storage: powder at 4°C to -20°C; in solvent store at -80°C for long-term stability.
  • Intended for research use only; not for human or clinical applications.

Catalog No. 50-002-96427 Supplier Medchemexpress LLC Supplier No. HYN058710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demethylzeylasteral is an orally active triterpenoid compound isolated from Tripterygium wilfordii that functions as an apoptosis inducer and has reported anti-inflammatory and antitumor activities. Supplied as a high-purity solid for research applications, it is suitable for use in biochemical and cell-based studies when prepared according to laboratory protocols.

  • Apoptosis inducer with reported anti-inflammatory and antitumor activity.
  • CAS number 107316-88-1 for unambiguous identification.
  • Molecular formula C29H36O6; molecular weight 480.59.
  • High purity (≈99.9%) in solid form, suitable for experimental work.
  • Available in multiple sizes, including 10 mg and millimolar DMSO solutions.
  • Storage: powder at 4°C to -20°C; in solvent store at -80°C for long-term stability.
  • Intended for research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.