Learn More
TARGETMOL CHEMICALS INC DEMETHYLZEYLASTERAL 25MG
SDP
Catalog No. 502576947
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Demethylzeylasteral (Demethylzeylasteral (T-96)) has strong immunosuppressive activity can be used in the fields of organ transplantation and autoimmune disorders. The risk of elevated serum concentrations of estradiol due to the inhibition of estradiol glucuronidation by Demethylzeylasteral. Demethylzeylasteral increases both activation and inactivation time constants of Ca(2+) currents can inhibit significantly the sperm acrosome reaction initiated by progesterone. purity: 99%
Catalog No. 50-257-6947 Supplier TARGETMOL CHEMICALS INC Supplier No. T341825MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Demethylzeylasteral (Demethylzeylasteral (T-96)) has strong immunosuppressive activity can be used in the fields of organ transplantation and autoimmune disorders. The risk of elevated serum concentrations of estradiol due to the inhibition of estradiol glucuronidation by Demethylzeylasteral. Demethylzeylasteral increases both activation and inactivation time constants of Ca(2+) currents can inhibit significantly the sperm acrosome reaction initiated by progesterone. purity: 99%