Learn More
Medchemexpress LLC HY-N0587 5mg Medchemexpress, Demethylzeylasteral CAS:107316-88-1 Purity:>98%
SDP
Catalog No. 501970292
Encompass_Preferred
Medchemexpress, HY-N0587 5mg Demethylzeylasteral CAS:107316-88-1 Demethylzeylasteral is a triterpene compound isolated from Tripterygium wilfordii Hook F, with anti-inflammatory, immunosuppressive and anti-tumor activities[3][4][5]. Demethylzeylasteral can significantly alleviates atherosclerosis (AS)[5]. Demethylzeylasteral inhibits triple-negative breast cancer invasion by blocking the canonical and non-canonical TGF-β signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0292 Supplier Medchemexpress LLC Supplier No. HYN05875MG
Medchemexpress, HY-N0587 5mg Demethylzeylasteral CAS:107316-88-1 Demethylzeylasteral is a triterpene compound isolated from Tripterygium wilfordii Hook F, with anti-inflammatory, immunosuppressive and anti-tumor activities[3][4][5]. Demethylzeylasteral can significantly alleviates atherosclerosis (AS)[5]. Demethylzeylasteral inhibits triple-negative breast cancer invasion by blocking the canonical and non-canonical TGF-β signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.