Learn More
Selleck Chemical LLC Demethylzeylasteral (T-96) 5mg 107316-88-1
SDP
Supplier:  Selleck Chemical LLC S36085MG
Encompass_Preferred
Demethylzeylasteral (T-96), the active component isolated from Tripterygium wilfordii Hook F., inhibits UDP-glucuronosyltransferase (UGT) isoforms UGT1A6 and UGT2B7 with immunosuppressive effects. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8397