Learn More
Crescent Chemical Co Inc DEMETON-S-METHYL-SULFONE 100MG
SDP
Supplier:  Crescent Chemical Co Inc PSC12144000
Encompass_Preferred
Demeton-S-methyl sulfone Insecticide (Metabolite neat) CAS 17040-19-6 Molecular Weight - 262.2841 Storage - -18°C Catalog Num - DRE-C12144000 chemical
Catalog No. 50-732-79