Learn More
MOLTOX Molecular Toxicology Inc Demi Frazer Broth, 225 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948822
Encompass_Preferred
Demi-Fraser Broth, in conjunction with Fraser Broth Supplement, is used to selectively and differentially enrich Listeria from food and environmental samples. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-822 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S027
Demi-Fraser Broth, in conjunction with Fraser Broth Supplement, is used to selectively and differentially enrich Listeria from food and environmental samples. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.