Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Demineralizer Cartridges
SCI_ED

Catalog No. S50915 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cartridge Holder
Cartridge, High-capacity Demineralizer
Cartridge, Ultrapure Demineralizer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S50915 Cartridge, High-capacity Demineralizer
S508171 Cartridge Holder
S50914 Cartridge, Ultrapure Demineralizer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S50915 Supplier Thermo Scientific™ Supplier No. D8901/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Special resin dye changes color when cartridge is exhausted.

Hose nipples on each end of these cartridges easily connect to your water supply with ordinary lab tubing (0.3 in. I.D. [9.5mm]). Resins in the deionization cartridge contain a special dye that changes color when the cartridge is exhausted.

High-Capacity Cartridge

  • Has a two-bed demineralizer with selected cation and anion resin beds
  • Delivers more liters per cartridge than mixed-bed types
  • Average resistance of effluent is 175kilohm·cm
  • Total ion-exchange capacity is 1725 grains (as NaCl)
UltraPure Cartridge
  • Produces high-resistance demineralized water from tap water
  • Increases purity of water from central demineralizer
  • Total ion-exchange capacity is 1200 grains (as NaCl), 849 grains (as NaCl) to 1 megohm·cm endpoint
  • pH is usually between 6.8 and 7.2
  • Water is largely free of silica and CO2

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 1725 grains as NaCl
Type Cartridge, High-capacity Demineralizer
For Use With (Application) Water purification
Diameter (Metric) 9.5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.