Learn More
Hach Company Demineralizer Cartridge, Organic Removal
Supplier:  Hach Company 2074200
Encompass
Activated carbon removes organic and chlorines from feed water. Place ahead of Ultra Pure or High Capacity cartridge. Replace after second demineralizer cartridge replacement.
Catalog No. 50-199-5229