Learn More
NIKON INSTRUMENTS INC DEMO HPZ4 WITH XEON
Catalog No. NC3315880
Encompass
  • Intel Xeon 4.6 GHz 20-core processor
  • 256 GB RAM, upgradeable to 512 GB
  • NVIDIA RTX A4000 16 GB GPU
  • 1 TB M.2 SSD for OS
  • 12 TB SATA data drive
  • Dual 10Gb Ethernet ports
  • 8x USB-A, 2x 20Gbps USB-C ports
  • DVD-RW optical drive
  • Windows 11 64-bit Professional OS
  • 27-inch 4K UHD display with 3840 x 2160 resolution
  • NIS-Elements software compatible
  • General analysis and AI modules included
  • 3-year HP warranty (parts, labor, onsite)
Catalog No. NC3315880 Supplier NIKON INSTRUMENTS INC Supplier No. 77019806
Add to Cart
Add to Cart
Request a Quote
  • Intel Xeon 4.6 GHz 20-core processor
  • 256 GB RAM, upgradeable to 512 GB
  • NVIDIA RTX A4000 16 GB GPU
  • 1 TB M.2 SSD for OS
  • 12 TB SATA data drive
  • Dual 10Gb Ethernet ports
  • 8x USB-A, 2x 20Gbps USB-C ports
  • DVD-RW optical drive
  • Windows 11 64-bit Professional OS
  • 27-inch 4K UHD display with 3840 x 2160 resolution
  • NIS-Elements software compatible
  • General analysis and AI modules included
  • 3-year HP warranty (parts, labor, onsite)