Learn More
Grainger Demo Screwdriver Set Slot/Phillips 7 pcs
Catalog No. 502764680
Encompass_Preferred
The WIHA Demolition Screwdriver Set is a 6-piece set of screwdrivers designed for heavy duty use. The set includes slotted, Phillips, Torx, hex and square screwdrivers with strike cap handles that provide extra grip and leverage. The blades are made from chrome vanadium steel for durability and feature a demagnetized treatment to avoid materials sticking. The sizes included are #2 Phillips, 1/4" and 5/16" slotted, T10 and T20 Torx, 1/4" and 5/16" square. This professional quality screwdriver set is ideal for demolition work, disassembly, prying, chiseling and scraping. The strike cap handles allow the user to tap or pound on the end.
Catalog No. 50-276-4680 Supplier Grainger Supplier No. 170G08
The WIHA Demolition Screwdriver Set is a 6-piece set of screwdrivers designed for heavy duty use. The set includes slotted, Phillips, Torx, hex and square screwdrivers with strike cap handles that provide extra grip and leverage. The blades are made from chrome vanadium steel for durability and feature a demagnetized treatment to avoid materials sticking. The sizes included are #2 Phillips, 1/4" and 5/16" slotted, T10 and T20 Torx, 1/4" and 5/16" square. This professional quality screwdriver set is ideal for demolition work, disassembly, prying, chiseling and scraping. The strike cap handles allow the user to tap or pound on the end.