Learn More
Grainger Demo Screwdriver Set Slot/Phillips 5 pcs
Catalog No. 502764743
Encompass_Preferred
The WIHA Demolition Screwdriver Set is a 5-piece set of screwdrivers designed for heavy duty use. The set includes slotted and Phillips head screwdrivers in sizes ranging from 5/16" to 1/4". The blades are made from chrome vanadium steel for durability and feature a demagnetized treatment to avoid materials sticking. The handles have a multi-component grip and strike cap for applying force. This professional quality screwdriver set is ideal for demolition work, chiseling, scraping, and prying. With German engineering and quality manufacturing, it provides strength and longevity. The set comes in a slim protective case for storage and portability.
Catalog No. 50-276-4743 Supplier Grainger Supplier No. 170G09
Add to Cart
Add to Cart
The WIHA Demolition Screwdriver Set is a 5-piece set of screwdrivers designed for heavy duty use. The set includes slotted and Phillips head screwdrivers in sizes ranging from 5/16" to 1/4". The blades are made from chrome vanadium steel for durability and feature a demagnetized treatment to avoid materials sticking. The handles have a multi-component grip and strike cap for applying force. This professional quality screwdriver set is ideal for demolition work, chiseling, scraping, and prying. With German engineering and quality manufacturing, it provides strength and longevity. The set comes in a slim protective case for storage and portability.